Malonaldehyde bis(phenylimine) monohydrochloride, 97+%, Thermo Scientific Chemicals
Malonaldehyde bis(phenylimine) monohydrochloride, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Malonaldehyde bis(phenylimine) monohydrochloride, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number B21721.22
also known as B21721-22
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
Add to cart
Malonaldehyde bis(phenylimine) monohydrochloride, 97+%, Thermo Scientific Chemicals
Catalog numberB21721.22
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS123071-42-1
IUPAC NameN1,N3-diphenylpropane-1,3-diimine hydrochloride
Molecular FormulaC15H15ClN2
InChI KeyFLMLOZWXXPLHIE-UHFFFAOYSA-N
SMILESCl.C(C=NC1=CC=CC=C1)C=NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
FormPowder
Assay (Titration ex Chloride)≥97.0 to ≤103.0%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤2.25%
Malonaldehyde bis(phenylimine) monohydrochloride is a useful fluorescent cyanine dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Malonaldehyde bis(phenylimine) monohydrochloride is a useful fluorescent cyanine dye.

Solubility
Soluble in water (partly).

Notes
Hygroscopic, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. F.S. Mair; D. Scully; A.J. Edwards; P.R. Raithby; R. Snaith. Synthetic, structural, spectroscopic and calculational studies of a lithium β-diketinimate complex. Polyhedron, 1995, 14 (17-18), 2397-2401.
  2. Kai Licha; Carsten Hessenius; Andreas Becker; Peter Henklein; Michael Bauer; Stefan Wisniewski; Bertram Wiedenmann; and Wolfhard Semmler . Synthesis, Characterization, and Biological Properties of Cyanine-Labeled Somatostatin Analogues as Receptor-Targeted Fluorescent Probes. Bioconjugate Chem, 2001, 12(1), 44-50.