Thermo Scientific Chemicals

3-Bromobiphenyl, 99%, Thermo Scientific Chemicals

Catalog number: B21726.06
5 g, Each
Thermo Scientific Chemicals

3-Bromobiphenyl, 99%, Thermo Scientific Chemicals

Catalog number: B21726.06
5 g, Each
Quantity
Catalog number: B21726.06
also known as B21726-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2113-57-7
IUPAC Name
3-bromo-1,1'-biphenyl
Molecular Formula
C12H9Br
InChI Key
USYQKCQEVBFJRP-UHFFFAOYSA-N
SMILES
BrC1=CC=CC(=C1)C1=CC=CC=C1
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥98.5%
Refractive Index
1.6395-1.6425 @ 20?C

Description

3-Bromobiphenyl is utilized as an electrochemical immunosensor based on poly(dopamine) coated gold nanocluster for brominated flame retardants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobiphenyl is utilized as an electrochemical immunosensor based on poly(dopamine) coated gold nanocluster for brominated flame retardants.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text