Thermo Scientific Chemicals

Rosolic acid, Thermo Scientific Chemicals

Catalog number: B21737.22
100 g, Each
Thermo Scientific Chemicals

Rosolic acid, Thermo Scientific Chemicals

Catalog number: B21737.22
100 g, Each
Quantity
Catalog number: B21737.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
603-45-2
IUPAC Name
4-[bis(4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
Molecular Formula
C19H14O3
InChI Key
FYEHYMARPSSOBO-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)=C1C=CC(=O)C=C1
Appearance (Color)
Dark red
Comment
Material sourced in the U.S. and in other countries
Loss on Drying
≤8%
Appearance (Color)
Dark red
Form
Powder

Description

Rosolic acid is used as a pH indicator in the range from 5.0 to 6.8. It acts as an intermediate in the manufacturing dyes. It is involved in the activation of heme oxygenase and in the inhibition of DNA fragmentation. It finds application in the selective isolation of coliform bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rosolic acid is used as a pH indicator in the range from 5.0 to 6.8. It acts as an intermediate in the manufacturing dyes. It is involved in the activation of heme oxygenase and in the inhibition of DNA fragmentation. It finds application in the selective isolation of coliform bacteria.

Solubility
Soluble in alcohol, sodium hydroxide, potassium hydroxide and strong acids. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text