Thermo Scientific Chemicals

trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: B21742.18
50 g, Each
Thermo Scientific Chemicals

trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: B21742.18
50 g, Each
Quantity
Catalog number: B21742.18
also known as B21742-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1197-18-8
IUPAC Name
4-(aminomethyl)cyclohexane-1-carboxylic acid
Molecular Formula
C8H15NO2
InChI Key
GYDJEQRTZSCIOI-UHFFFAOYSA-N
SMILES
NCC1CCC(CC1)C(O)=O
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

It is an antifibrinolytic agent that is found to inhibit plasmin-induced fibrinolysis. Used in various hemorrhagic diseases , abnormal bleeding in operations. It is also used as a lysine analogue to characterize binding sites in plasminogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an antifibrinolytic agent that is found to inhibit plasmin-induced fibrinolysis. Used in various hemorrhagic diseases , abnormal bleeding in operations. It is also used as a lysine analogue to characterize binding sites in plasminogen.

Solubility
Soluble in water (167 mg/ml), ethanol (<1mg/mL at 25°C). Very slightly soluble in ether, DMSO (<1mg/mL at 25°C), and alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text