Thermo Scientific Chemicals

Dimethyl sulfone, 98%, Thermo Scientific Chemicals

Catalog number: B21747.30
250 g, Each
Thermo Scientific Chemicals

Dimethyl sulfone, 98%, Thermo Scientific Chemicals

Catalog number: B21747.30
250 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B21747.30
also known as B21747-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-71-0
IUPAC Name
methanesulfonylmethane
Molecular Formula
C2H6O2S
InChI Key
HHVIBTZHLRERCL-UHFFFAOYSA-N
SMILES
CS(C)(=O)=O
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
106.0-113.0?C
Appearance (Color)
White
Assay (GC)
≥97.5%

Description

Dimethyl sulfone is used as a high-temperature solvent for both inorganic and organic substances. It acts as an intermediate as well as medium in organic synthesis. It plays an important role as a carbon source by the airborne bacteria Afipia. It is also employed as an extraction solvent for the separation of aromatic compounds like benzene, toluene and xylenes from aliphatic hydrocarbons. Further, it is used in the polymerization process. It finds application in pharmaceuticals, agrochemicals, paint and coating materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl sulfone is used as a high-temperature solvent for both inorganic and organic substances. It acts as an intermediate as well as medium in organic synthesis. It plays an important role as a carbon source by the airborne bacteria Afipia. It is also employed as an extraction solvent for the separation of aromatic compounds like benzene, toluene and xylenes from aliphatic hydrocarbons. Further, it is used in the polymerization process. It finds application in pharmaceuticals, agrochemicals, paint and coating materials.

Solubility
Soluble in water, methanol, ethanol and acetone.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text