4-Nitrobenzylamine hydrochloride, 94%, Thermo Scientific Chemicals
4-Nitrobenzylamine hydrochloride, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrobenzylamine hydrochloride, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number B21748.14
also known as B21748-14
Price (USD)/ Each
322.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
322.00
Add to cart
4-Nitrobenzylamine hydrochloride, 94%, Thermo Scientific Chemicals
Catalog numberB21748.14
Price (USD)/ Each
322.00
-
Add to cart
Chemical Identifiers
CAS18600-42-5
IUPAC Name(4-nitrophenyl)methanaminium chloride
Molecular FormulaC7H9ClN2O2
InChI KeySMIXZZMSWYOQPW-UHFFFAOYSA-N
SMILES[Cl-].[NH3+]CC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow to brown
Assay (Titration ex Chloride)≥92.0 to ≤108.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only