Thermo Scientific Chemicals

Homomorpholine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: B21749.06
5 g, Each
Thermo Scientific Chemicals

Homomorpholine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: B21749.06
5 g, Each
Quantity
Catalog number: B21749.06
also known as B21749-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
178312-62-4
IUPAC Name
1,4-oxazepane hydrochloride
Molecular Formula
C5H12ClNO
InChI Key
LJRCWNIWOVZLKS-UHFFFAOYSA-N
SMILES
Cl.C1CNCCOC1
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Appearance (Color)
Pale orange to brown
Water Content (Karl Fischer Titration)
≤1.0%

Description

Homomorpholine Hydrochloride is a useful synthetic intermediate. It is used to prepare homomorpholine oxazolidinone with antibacterial activities. It is also used to synthesize A2A adenosine receptor antagonists for the treatment of Parkinson's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Keep away from oxidizing agents, water/moisture. Store under inert atmosphere. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text