4-Chloro-2-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals
4-Chloro-2-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloro-2-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B21755.14
also known as B21755-14
Price (USD)/ Each
41.20
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
41.20
Add to cart
4-Chloro-2-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals
Catalog numberB21755.14
Price (USD)/ Each
41.20
-
Add to cart
Chemical Identifiers
CAS445-03-4
IUPAC Name4-chloro-2-(trifluoromethyl)aniline
Molecular FormulaC7H5ClF3N
InChI KeyCVINWVPRKDIGLL-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=C1C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5045-1.5095 @ 20°C
Appearance (Color)Clear colorless to yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only