Thermo Scientific Chemicals

2-Benzyl-2-imidazoline hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: B21764.14
25 g, Each
Thermo Scientific Chemicals

2-Benzyl-2-imidazoline hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: B21764.14
25 g, Each
Quantity
Catalog number: B21764.14
also known as B21764-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59-97-2
IUPAC Name
hydrogen 2-benzyl-4,5-dihydro-1H-imidazole chloride
Molecular Formula
C10H13ClN2
InChI Key
RHTNTTODYGNRSP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].C(C1=NCCN1)C1=CC=CC=C1
Appearance (Color)
White to cream to yellow
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Melting Point (clear melt)
170.0-176.0?C

Description

An alpha adrenergic blocking agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text