Thermo Scientific Chemicals

Glycidyl phenyl ether, 99%, Thermo Scientific Chemicals

Catalog number: B21768.0B
1000 g, Each
Thermo Scientific Chemicals

Glycidyl phenyl ether, 99%, Thermo Scientific Chemicals

Catalog number: B21768.0B
1000 g, Each
Quantity
Catalog number: B21768.0B
also known as B21768-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122-60-1
IUPAC Name
2-(phenoxymethyl)oxirane
Molecular Formula
C9H10O2
InChI Key
FQYUMYWMJTYZTK-UHFFFAOYNA-N
SMILES
C(OC1=CC=CC=C1)C1CO1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.5295-1.5325 @ 20?C

Description

2-(Phenoxymethyl)oxirane is used in the synthesis of hydrazide-containing inhibitors of HIV-1 integrase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Phenoxymethyl)oxirane is used in the synthesis of hydrazide-containing inhibitors of HIV-1 integrase.

Solubility
Soluble in water (2.4g/L).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text