Thermo Scientific Chemicals

(S)-(+)-1,2-Propanediol, 97%, Thermo Scientific Chemicals

Catalog number: B21773.14
25 g, Each
Thermo Scientific Chemicals

(S)-(+)-1,2-Propanediol, 97%, Thermo Scientific Chemicals

Catalog number: B21773.14
25 g, Each
Quantity
Catalog number: B21773.14
also known as B21773-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4254-15-3
IUPAC Name
propane-1,2-diol
Molecular Formula
C3H8O2
InChI Key
DNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES
CC(O)CO
Appearance (Color)
Clear colorless
Form
Liquid or viscous liquid
Assay (GC)
≥96.0%
Refractive Index
1.4305-1.4355 @ 20?C
Optical Rotation
+15.0? to +17.0? (neat)

Description

(S)-(+)-1,2-Propanediol acts as an organic solvent and diluent used in pharmaceutical preparations. It is also used as a chiral synthetic intermediate and substrate for enzyme studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-1,2-Propanediol acts as an organic solvent and diluent used in pharmaceutical preparations. It is also used as a chiral synthetic intermediate and substrate for enzyme studies.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides, oxidizing agents,chloroformates and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text