Trifluoromethanesulfonyl chloride, 98%, Thermo Scientific Chemicals
Trifluoromethanesulfonyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trifluoromethanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B21774.14
also known as B21774-14
Price (USD)/ Each
212.65
Online exclusive
236.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
212.65
Online exclusive
236.00 
Save 23.35 (10%)
Add to cart
Trifluoromethanesulfonyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberB21774.14
Price (USD)/ Each
212.65
Online exclusive
236.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS421-83-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear or slightly turbid, colorless to yellow
Assay (GC)>97.5%
Refractive Index1.3310-1.3370 @ 20?C
FormLiquid
Ru(II) complex-catalyzed trifluoromethylating agent for aromatics and alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ru(II) complex-catalyzed trifluoromethylating agent for aromatics and alkenes.

Solubility
Hydrolyzes with water.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Nobumasa Kamigata; Takeshi Ohtsuka; Takamasa Fukushima; Masato Yoshida; and Toshio Shimizu. Direct perfluoroalkylation of aromatic and heteroaromatic compounds with perfluoroalkanesulfonyl chlorides catalyzed by a ruthenium(II) phosphine complex.J. Chem. Soc., Perkin Trans. 1.19941339-1346.
  2. Nobumasa Kamigata;Takamasa Fukushima; Yoshie Terakawa; Masato Yoshida; and Hideo Sawada. Novel perfluoroalkylation of alkenes with perfluoroalkanesulphonyl chlorides catalyzed by a ruthenium(II) complex.J. Chem. Soc., Perkin Trans. 1.1991(3), 627-633.