Desyl chloride, 98%, Thermo Scientific Chemicals
Desyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Desyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B21778.14
also known as B21778-14
Price (USD)/ Each
107.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
107.00
Add to cart
Desyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberB21778.14
Price (USD)/ Each
107.00
-
Add to cart
Chemical Identifiers
CAS447-31-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (GC)≥97.5%
Melting Point (clear melt)63.0-69.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only