Thermo Scientific Chemicals

2,6-Dimethoxy-4-methylphenol, 97%, Thermo Scientific Chemicals

Catalog number: B21801.22
100 g, Each
Thermo Scientific Chemicals

2,6-Dimethoxy-4-methylphenol, 97%, Thermo Scientific Chemicals

Catalog number: B21801.22
100 g, Each
Quantity
Catalog number: B21801.22
also known as B21801-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6638-05-7
IUPAC Name
2,6-dimethoxy-4-methylphenol
Molecular Formula
C9H12O3
InChI Key
ZFBNNSOJNZBLLS-UHFFFAOYSA-N
SMILES
COC1=CC(C)=CC(OC)=C1O
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
34.0-44.0?C
Assay (GC)
≥96.0%
Appearance (Color)
Pale cream to cream to yellow to pale brown

Description

Used as a flavor and fragrance agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavor and fragrance agents.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text