Thermo Scientific Chemicals

6-Bromo-1-hexanol, 96%, Thermo Scientific Chemicals

Catalog number: B21803.03
1 g, Each
Thermo Scientific Chemicals

6-Bromo-1-hexanol, 96%, Thermo Scientific Chemicals

Catalog number: B21803.03
1 g, Each
Quantity
Catalog number: B21803.03
also known as B21803-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4286-55-9
IUPAC Name
6-bromohexan-1-ol
Molecular Formula
C6H13BrO
InChI Key
FCMCSZXRVWDVAW-UHFFFAOYSA-N
SMILES
OCCCCCCBr
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥95.0%
Refractive Index
1.4790-1.4840 @ 20?C
Form
Liquid

Description

A product for proteomics research. Also used as a reagent in the preparation of nitric oxide-donating analogues of Sulindac (S699215), a non-steroidal anti-inflammatory drug that has the ability to prevent colon cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A product for proteomics research. Also used as a reagent in the preparation of nitric oxide-donating analogues of Sulindac (S699215), a non-steroidal anti-inflammatory drug that has the ability to prevent colon cancer.

Solubility
Soluble in chloroform and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text