Reactant used for preparation of fluorescent probes for imaging of tumors, hypoxia-sensitive fluorescent probes for real-time imaging of acute ischemia and near IR fluorescent probe for imaging integrin receptor expression.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reactant used for preparation of fluorescent probes for imaging of tumors , hypoxia-sensitive fluorescent probes for real-time imaging of acute ischemia and near IR fluorescent probe for imaging integrin receptor expression.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
Koji Miki et. al. High-contrast fluorescence imaging of tumors in vivo using nanoparticles of amphiphilic brush-like copolymers produced by ROMP. Angewandte Chemie. International edition in English., 2011, 50 (29), 6567-6570.
Kazuki Kiyose et. al. Hypoxia-sensitive fluorescent probes for in vivo real-time fluorescence imaging of acute ischemia. Journal of the American Chemical Society, 2010, 132 (45), 15846-15848.