Thermo Scientific Chemicals

1,1,2-Trimethyl-1H-benzo[e]indole, 97%, Thermo Scientific Chemicals

Catalog number: B21804.14
25 g, Each
Thermo Scientific Chemicals

1,1,2-Trimethyl-1H-benzo[e]indole, 97%, Thermo Scientific Chemicals

Catalog number: B21804.14
25 g, Each
Quantity
Catalog number: B21804.14
also known as B21804-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
41532-84-7
IUPAC Name
1,1,2-trimethyl-1H-benzo[e]indole
Molecular Formula
C15H15N
InChI Key
WJZSZXCWMATYFX-UHFFFAOYSA-N
SMILES
CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C
Appearance (Color)
Yellow to brown
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
112.0-121.0?C

Description

Reactant used for preparation of fluorescent probes for imaging of tumors, hypoxia-sensitive fluorescent probes for real-time imaging of acute ischemia and near IR fluorescent probe for imaging integrin receptor expression.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant used for preparation of fluorescent probes for imaging of tumors , hypoxia-sensitive fluorescent probes for real-time imaging of acute ischemia and near IR fluorescent probe for imaging integrin receptor expression.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text