Thermo Scientific Chemicals

Vinyl benzoate, 95%, Thermo Scientific Chemicals

Catalog number: B21805.36
500 g, Each
Thermo Scientific Chemicals

Vinyl benzoate, 95%, Thermo Scientific Chemicals

Catalog number: B21805.36
500 g, Each
Quantity
Catalog number: B21805.36
also known as B21805-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
769-78-8
IUPAC Name
ethenyl benzoate
Molecular Formula
C9H8O2
InChI Key
KOZCZZVUFDCZGG-UHFFFAOYSA-N
SMILES
C=COC(=O)C1=CC=CC=C1
Appearance (Color)
Clear colorless
Assay (GC)
≥94.0%
Solution Test
10% in methanol: Clear
Solution Test
10% in diethyl ether: Clear
Form
Liquid

Description

It is used as chemical reagent and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as chemical reagent and pharmaceutical intermediates.

Solubility
Soluble in methanol

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text