Thermo Scientific Chemicals

Indole-5-carbonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: B21810.03
1 g, Each
Thermo Scientific Chemicals

Indole-5-carbonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: B21810.03
1 g, Each
Quantity
Catalog number: B21810.03
also known as B21810-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15861-24-2
IUPAC Name
1H-indole-5-carbonitrile
Molecular Formula
C9H6N2
InChI Key
YHYLDEVWYOFIJK-UHFFFAOYSA-N
SMILES
N#CC1=CC=C2NC=CC2=C1
Form
Powder
Assay (GC)
≥98.0%
Melting Point (clear melt)
103.0-109.0?C
Appearance (Color)
White to cream to yellow

Description

Reactant used for parallel synthesis of dihydroisoquinolines via silver and L-proline co-catalyzed three-component coupling reaction, preparation of novel PPARα/γ dual agonists for potential treatment of metabolic syndrome and IDDM, chemo selective and regioselective preparation of benzoyl indoles, 4,5-dihydrocyclopenta[c]quinolines by palladium-catalyzed ring-expansion reaction alkynes, using O2 as the oxidant and vinylindoles by hydroarylation of alkynes using indium bromide catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant used for parallel synthesis of dihydroisoquinolines via silver and L-proline co-catalyzed three-component coupling reaction, preparation of novel PPARα/γ dual agonists for potential treatment of metabolic syndrome and IDDM, chemo selective and regioselective preparation of benzoyl indoles, 4,5-dihydrocyclopenta[c]quinolines by palladium-catalyzed ring-expansion reaction alkynes, using O2 as the oxidant and vinylindoles by hydroarylation of alkynes using indium bromide catalyst.

Solubility
Soluble in chloroform, hexane and methanol. Insoluble in water.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text