Thermo Scientific Chemicals

Ethanesulfonic acid sodium salt, 98%, may cont. ca 2% water, Thermo Scientific Chemicals

Catalog number: B21824.06
5 g, Each
Thermo Scientific Chemicals

Ethanesulfonic acid sodium salt, 98%, may cont. ca 2% water, Thermo Scientific Chemicals

Catalog number: B21824.06
5 g, Each
Quantity
Catalog number: B21824.06
also known as B21824-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5324-47-0
IUPAC Name
sodium ethanesulfonate
Molecular Formula
C2H5NaO3S
InChI Key
KQFAFFYKLIBKDE-UHFFFAOYSA-M
SMILES
[Na+].CCS([O-])(=O)=O
Appearance (Color)
White
Assay (ex Sulphated Ash)
≥97.5 to ≤102.5%
Form
Powder
Water Content (Karl Fischer Titration)
≤2.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text