2-Ethylacrolein, tech. 90%, stab. with 50ppm hydroquinone, Thermo Scientific Chemicals
2-Ethylacrolein, tech. 90%, stab. with 50ppm hydroquinone, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Ethylacrolein, tech. 90%, stab. with 50ppm hydroquinone, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B21839.14
also known as B21839-14
Price (USD)/ Each
236.65
Online exclusive
263.00 
Save 26.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
236.65
Online exclusive
263.00 
Save 26.35 (10%)
Add to cart
2-Ethylacrolein, tech. 90%, stab. with 50ppm hydroquinone, Thermo Scientific Chemicals
Catalog numberB21839.14
Price (USD)/ Each
236.65
Online exclusive
263.00 
Save 26.35 (10%)
-
Add to cart
Chemical Identifiers
CAS922-63-4
IUPAC Name2-methylidenebutanal
Molecular FormulaC5H8O
InChI KeyGMLDCZYTIPCVMO-UHFFFAOYSA-N
SMILESCCC(=C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormLiquid
Assay (GC)>88.0%
Water Content (Karl Fischer Titration)<1.0%
Refractive Index1.4180-1.4270 @ 20?C
2-Ethylacrolein has been used in total synthesis of (±)-tabersonine, Aspidosperma alkaloid and synthesis of conjugate with bovine serum albumin tethered to Tn.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylacrolein has been used in total synthesis of (+/-)-tabersonine, Aspidosperma alkaloid and synthesis of conjugate with bovine serum albumin tethered to Tn.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store at 2-8°C.
RUO – Research Use Only

General References:

  1. Tilman Neudecker.; Erwin Eder.; Christoph Deininger.; Dietrich Henschler. Mutagenicity of 2-methylacrolein, 2-ethylacrolein and 2-propylacrolein in Salmonella typhimurium TA 100. A comparative study.Mutation Research Letters. 1991, 264 (4),193-196.
  2. D.F Parra.;L.P Mercuri.; J.R Matos.; H.F Brito.; R.R Romano. Thermal behavior of the epoxy and polyester powder coatings using thermogravimetry/differential thermal analysis coupled gas chromatography/mass spectrometry (TG/DTA-GC/MS) technique: identification of the degradation products.Thermochim. Acta. 2002, 386 (22), 143-151.