11-Bromo-1-undecene, 90+%, Thermo Scientific Chemicals
11-Bromo-1-undecene, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

11-Bromo-1-undecene, 90+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21849.06
also known as B21849-06
Price (USD)/ Each
80.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
80.90
Add to cart
11-Bromo-1-undecene, 90+%, Thermo Scientific Chemicals
Catalog numberB21849.06
Price (USD)/ Each
80.90
-
Add to cart
Chemical Identifiers
CAS7766-50-9
IUPAC Name11-bromoundec-1-ene
Molecular FormulaC11H21Br
InChI KeyYPLVPFUSXYSHJD-UHFFFAOYSA-N
SMILESBrCCCCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥90.0%
Refractive Index1.4650-1.4710 @ 20?C
FormLiquid
Identification (FTIR)Conforms
11-Bromo-1-undecene is used in advanced polymer dielectrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
11-Bromo-1-undecene is used in advanced polymer dielectrics.

Solubility
Soluble in water, 0.2422 mg/L @ 25°C (est).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Oĝuz Türünç; Lucas Montero de Espinosa1 and Michael A. R. Meier. TRenewable Polyethylene Mimics Derived from Castor Oil. Macromolecular Rapid Communications. 2011, 32 (17), 1357-1361.
  2. Laura J. Yeager; Dafni G. Amirsakis; Elan Newman; Robin L. Garrell. Synthesis of epoxy-terminated dialkyl disulfides for polymerizable self-assembled monolayers. Tetrahedron Letters. 1998, 39 (46), 8409-8412.