Thermo Scientific Chemicals

Methyl 2-methoxy-5-nitrobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: B21863.06
5 g, Each
Thermo Scientific Chemicals

Methyl 2-methoxy-5-nitrobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: B21863.06
5 g, Each
Quantity
Catalog number: B21863.06
also known as B21863-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34841-11-7
IUPAC Name
methyl 2-methoxy-5-nitrobenzoate
Molecular Formula
C9H9NO5
InChI Key
DOBFJVVTBNTGCW-UHFFFAOYSA-N
SMILES
COC(=O)C1=C(OC)C=CC(=C1)[N+]([O-])=O
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
98.0-104.0?C
Appearance (Color)
White to pale cream to pale yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text