Florisil∣r, 100-200 mesh, Thermo Scientific Chemicals
Florisil∣r, 100-200 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Florisil∣r, 100-200 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number B21870.30
also known as B21870-30
Price (USD)/ Each
124.65
Online exclusive
138.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
124.65
Online exclusive
138.00 
Save 13.35 (10%)
Add to cart
Florisil∣r, 100-200 mesh, Thermo Scientific Chemicals
Catalog numberB21870.30
Price (USD)/ Each
124.65
Online exclusive
138.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1343-88-0
IUPAC Namesilicon(4+) magnesium(2+) trioxidandiide
Molecular FormulaMgO3Si
InChI KeyZADYMNAVLSWLEQ-UHFFFAOYSA-N
Molecular Weight (g/mol)100.39
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Magnesium oxide : 15.0-16.0%
Assay (unspecified)Silicon dioxide : 83.5-84.5%
Assay (unspecified)Sodium Sulfate : <1.0%
CommentActivated Magnesium silicate.
CommentFlorisil® is a registered trademark of U.S. Silica Company.
View more

Florisil is employed for polyether adsorbent, which has applications of adsorption and decoloring, adsorbent in gas chromatography, in vitamin analysis, antibiotic processing, anti caking agent, used in cosmetics as talcum & baby powder, general modified plastics, mid-range insulation materials, EVA shoes material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Florisil is employed for polyether adsorbent, which has applications of adsorption and decoloring, adsorbent in gas chromatography, in vitamin analysis, antibiotic processing, anti caking agent, used in cosmetics as talcum & baby powder, general modified plastics, mid-range insulation materials, EVA shoes material.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents, hydrogen fluoride.
RUO – Research Use Only

General References:

  1. Yongqiang Wang Dr.; Guozhong Wang Prof; Hongqiang Wang Dr.; Changhao Liang; Weiping Cai and Lide Zhang. Chemical-Template Synthesis of Microanoscale Magnesium Silicate Hollow Spheres for Waste-Water Treatment. Chem. Eur. J. 2010, 16 (11), 3497-3503.
  2. K. K. Carroll. Acid-treated florisil as an adsorbent for column chromatography. Journal of the American Oil Chemists Society. 1963, 40 (9), 413-419.