Thermo Scientific Chemicals

1,3-Diacetylbenzene, 97%, Thermo Scientific Chemicals

Catalog number: B21880.06
5 g, Each
Thermo Scientific Chemicals

1,3-Diacetylbenzene, 97%, Thermo Scientific Chemicals

Catalog number: B21880.06
5 g, Each
Quantity
Catalog number: B21880.06
also known as B21880-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6781-42-6
Assay (GC)
≥96.0%
Melting Point (clear melt)
29.0-38.0?C
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder or fused solid

Description

1,3-Diacetylbenzene was used in the preparation of polyhydroxylated analogs. It is an important raw material and intermediates in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diacetylbenzene was used in the preparation of polyhydroxylated analogs. It is an important raw material and intermediates in organic synthesis.

Solubility
Soluble in ethanol, benzene, chloroform.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text