Thermo Scientific Chemicals

3-Phenylazo-2,6-diaminopyridine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: B21886.06
5 g, Each
Thermo Scientific Chemicals

3-Phenylazo-2,6-diaminopyridine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: B21886.06
5 g, Each
Quantity
Catalog number: B21886.06
also known as B21886-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
136-40-3
IUPAC Name
hydrogen 3-(2-phenyldiazen-1-yl)pyridine-2,6-diamine chloride
Molecular Formula
C11H12ClN5
InChI Key
QQBPIHBUCMDKFG-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC1=CC=C(N=NC2=CC=CC=C2)C(N)=N1
Appearance (Color)
Red or purple
Form
powder
Assay (Titration ex Chloride)
>98.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text