3-Phenylazo-2,6-diaminopyridine hydrochloride, 98+%, Thermo Scientific Chemicals
3-Phenylazo-2,6-diaminopyridine hydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Phenylazo-2,6-diaminopyridine hydrochloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B21886.14
also known as B21886-14
Price (USD)/ Each
282.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
282.00
Add to cart
3-Phenylazo-2,6-diaminopyridine hydrochloride, 98+%, Thermo Scientific Chemicals
Catalog numberB21886.14
Price (USD)/ Each
282.00
-
Add to cart
Chemical Identifiers
CAS136-40-3
IUPAC Namehydrogen 3-(2-phenyldiazen-1-yl)pyridine-2,6-diamine chloride
Molecular FormulaC11H12ClN5
InChI KeyQQBPIHBUCMDKFG-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC1=CC=C(N=NC2=CC=CC=C2)C(N)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red or purple
Formpowder
Assay (Titration ex Chloride)>98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only