Thermo Scientific Chemicals

6-Methylquinoline, 98%, Thermo Scientific Chemicals

Catalog number: B21892.14
25 g, Each
Thermo Scientific Chemicals

6-Methylquinoline, 98%, Thermo Scientific Chemicals

Catalog number: B21892.14
25 g, Each
Quantity
Catalog number: B21892.14
also known as B21892-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91-62-3
Appearance (Color)
Clear colorless to yellow to pale brown
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.6120-1.6160 @ 20?C

Description

6-Methylquinoline can be used as primary carbon source in culture of Pseudomonas putida QP1. 6-Methylquinoline was used in the synthesis of fluorescent halide-sensitive quinolinium dyes and fluorescent probes for determination of chloride in biological systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methylquinoline can be used as primary carbon source in culture of Pseudomonas putida QP1. 6-Methylquinoline was used in the synthesis of fluorescent halide-sensitive quinolinium dyes and fluorescent probes for determination of chloride in biological systems.

Solubility
Not miscible or difficult to mix with water.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text