Thermo Scientific Chemicals

2-Amino-4-fluorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: B21907.03
1 g, Each
Thermo Scientific Chemicals

2-Amino-4-fluorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: B21907.03
1 g, Each
Quantity
Catalog number: B21907.03
also known as B21907-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
446-32-2
IUPAC Name
2-amino-4-fluorobenzoate
Molecular Formula
C7H5FNO2
InChI Key
LGPVTNAJFDUWLF-UHFFFAOYSA-M
SMILES
NC1=CC(F)=CC=C1C([O-])=O
Appearance (Color)
White to cream to pale brown
Form
Powder or chunks
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
187.0-196.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text