CHAPS, 98+%, Thermo Scientific Chemicals
CHAPS, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

CHAPS, 98+%, Thermo Scientific Chemicals

CHAPS, >-98%, C32H58N2O7S, CAS Number-75621-03-3, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, 25g, 614.88, H315-H319-H335, GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number B21927.14
Price (USD)/ Each
741.65
Online exclusive
824.00 
Save 82.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
741.65
Online exclusive
824.00 
Save 82.35 (10%)
Add to cart
CHAPS, 98+%, Thermo Scientific Chemicals
Catalog numberB21927.14
Price (USD)/ Each
741.65
Online exclusive
824.00 
Save 82.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75621-03-3
IUPAC Name3-({3-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]propyl}dimethylazaniumyl)propane-1-sulfonate
Molecular FormulaC32H58N2O7S
InChI KeyUMCMPZBLKLEWAF-BCTGSCMUSA-N
SMILESC[C@H](CCC(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay from Supplier's CofA≥98.0%
Conductivity≤10µS/cm (0.1M in water)
Identification (FTIR)Conforms
View more
Buffer; zwitterionic nondenaturing detergent for solubilizing membrane proteinsCHAPS is used in the laboratory to dissolve biological macromolecules such as proteins. It acts as a non-denaturing solvent used in purification of membrane proteins. It is also useful in conjunction with nonionic detergents like Triton X-100. Further, it is used as a non-denaturing zwitterionic detergent for membrane biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Buffer; zwitterionic nondenaturing detergent for solubilizing membrane proteinsCHAPS is used in the laboratory to dissolve biological macromolecules such as proteins. It acts as a non-denaturing solvent used in purification of membrane proteins. It is also useful in conjunction with nonionic detergents like Triton X-100. Further, it is used as a non-denaturing zwitterionic detergent for membrane biochemistry.

Solubility
Slightly soluble in water.

Notes
Store in a cool. Incompatible with oxidizing agent, strong acid and bases.
RUO – Research Use Only

General References:

  1. Rodi, P. M.; Gianello, M. D. B.; Corregido, M. C.; Gennaro, A. M. Comparative study of the interaction of CHAPS and Triton X-100 with the erythrocyte membrane. Biochim. Biophys. Acta 2014, 1838 (3), 859-866.
  2. Ghosh, A.; Saha, R.; Saha, B. Effect of CHAPS and CPC micelles on Ir(III) catalyzed Ce(IV) oxidation of aliphatic alcohols at room temperature and pressure. J. Mol. Liq. 2014, 196, 223-237.