Thermo Scientific Chemicals

Crystal Violet

Catalog number: B21932.36
500 g, Each
Thermo Scientific Chemicals

Crystal Violet

Catalog number: B21932.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B21932.36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
548-62-9
IUPAC Name
tris[4-(dimethylamino)phenyl]methylium chloride
Molecular Formula
C25H30ClN3
InChI Key
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
SMILES
[Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
Appearance (Color)
Dark green
Form
Crystals and/or crystalline powder
Assay (Dye)
≥96% (dry wt. basis, spectrophotometric)
Insoluble material
Ethanol : ≤0.5%
Loss on Drying
<9.0% (105?/constant wt.)

Description

Crystal Violet is used as an acid-base indicator, alcohol denaturant, biological stain, textile dye, and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Crystal Violet is used as an acid-base indicator, alcohol denaturant, biological stain, textile dye, and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.

Solubility
Miscible with water and chloroform. Partially miscible with alcohol and glycerol.

Notes
Incompatible with acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents and peroxides.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text