Thermo Scientific Chemicals

(R)-(-)-1,2-Propanediol, 98%, Thermo Scientific Chemicals

Catalog number: B21937.03
1 g, Each
Thermo Scientific Chemicals

(R)-(-)-1,2-Propanediol, 98%, Thermo Scientific Chemicals

Catalog number: B21937.03
1 g, Each
Quantity
Catalog number: B21937.03
also known as B21937-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4254-14-2
IUPAC Name
(2R)-propane-1,2-diol
Molecular Formula
C3H8O2
InChI Key
DNIAPMSPPWPWGF-GSVOUGTGSA-N
SMILES
C[C@@H](O)CO
Refractive Index
1.4310-1.4350 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid or viscous liquid
Assay (GC)
≥97.5%
Optical Rotation
-16.8 ± 0.1? (neat)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text