Thermo Scientific Chemicals

7-Diethylamino-4-methylcoumarin, 99%, Thermo Scientific Chemicals

Catalog number: B21938.36
500 g, Each
Thermo Scientific Chemicals

7-Diethylamino-4-methylcoumarin, 99%, Thermo Scientific Chemicals

Catalog number: B21938.36
500 g, Each
Quantity
Catalog number: B21938.36
also known as B21938-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91-44-1
IUPAC Name
7-(diethylamino)-4-methyl-2H-chromen-2-one
Molecular Formula
C14H17NO2
InChI Key
AFYCEAFSNDLKSX-UHFFFAOYSA-N
SMILES
CCN(CC)C1=CC=C2C(C)=CC(=O)OC2=C1
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
69.0-75.0?C
Appearance (Color)
White to pale cream

Description

7-(Diethylamino)-4-methylcoumarin, is potentially useful as a laser dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-(Diethylamino)-4-methylcoumarin, is potentially useful as a laser dye.

Solubility
Soluble in water, aqueous acid solutions, resins, varnishes, vinyls, ether, acetone, ordinary organic solvents, and alcohol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text