Thermo Scientific Chemicals

Sodium 1-tetradecyl sulfate, 95%, cont. up to ca 5% sodium methyl sulfate, Thermo Scientific Chemicals

Catalog number: B21941.03
1 g, Each
Thermo Scientific Chemicals

Sodium 1-tetradecyl sulfate, 95%, cont. up to ca 5% sodium methyl sulfate, Thermo Scientific Chemicals

Catalog number: B21941.03
1 g, Each
Quantity
Catalog number: B21941.03
also known as B21941-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1191-50-0
IUPAC Name
sodium tetradecyl sulfate
Molecular Formula
C14H29NaO4S
InChI Key
UPUIQOIQVMNQAP-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O
Appearance (Color)
White
Form
Powder
Assay from Supplier's CofA
≥94.0%

Description

Sodium 1-tetradecyl sulfate is used as an anionic surfactant in nickel plating bath, pickling, electrolytic cleaning, welding, leather dyeing and rubles polishes. It is used to study the time course of liquid drainage and foam coarsening and to investigate the effects of sclerosant foam preparation. It is used for the treatment of varicose and spider veins of the leg.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-tetradecyl sulfate is used as an anionic surfactant in nickel plating bath, pickling, electrolytic cleaning, welding, leather dyeing and rubles polishes. It is used to study the time course of liquid drainage and foam coarsening and to investigate the effects of sclerosant foam preparation. It is used for the treatment of varicose and spider veins of the leg.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text