Thermo Scientific Chemicals

2,3-Diphenyl-2-cyclopropen-1-one, 99% (dry wt.), may cont up to 5% water, Thermo Scientific Chemicals

Catalog number: B21954.14
25 g, Each
Thermo Scientific Chemicals

2,3-Diphenyl-2-cyclopropen-1-one, 99% (dry wt.), may cont up to 5% water, Thermo Scientific Chemicals

Catalog number: B21954.14
25 g, Each
Quantity
Catalog number: B21954.14
also known as B21954-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
886-38-4
IUPAC Name
diphenylcycloprop-2-en-1-one
Molecular Formula
C15H10O
InChI Key
HCIBTBXNLVOFER-UHFFFAOYSA-N
SMILES
O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream to pale brown or pale orange
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text