Thermo Scientific Chemicals

4-Chloro-3,5-dimethylphenol, 98+%, Thermo Scientific Chemicals

Catalog number: B21964.36
500 g, Each
Thermo Scientific Chemicals

4-Chloro-3,5-dimethylphenol, 98+%, Thermo Scientific Chemicals

Catalog number: B21964.36
500 g, Each
Quantity
Catalog number: B21964.36
also known as B21964-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-04-0
IUPAC Name
4-chloro-3,5-dimethylphenol
Molecular Formula
C8H9ClO
InChI Key
OSDLLIBGSJNGJE-UHFFFAOYSA-N
SMILES
CC1=CC(O)=CC(C)=C1Cl
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.0%
Melting Point (clear melt)
112.0-118.0?C
Appearance (Color)
White to pale cream

Description

It is used in hospitals and households for disinfection and sanitation. It is used in wound-cleansing applications and household antiseptics such as Dettol liquid, cream and ointments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in hospitals and households for disinfection and sanitation. It is used in wound-cleansing applications and household antiseptics such as Dettol liquid, cream and ointments.

Solubility
Soluble in benzene, toluene, glycerin, chloroform, acetone, isopropanol, alcohol, ether, terpenes and in solutions of alkali hydroxides. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text