Thermo Scientific Chemicals

3-Mercapto-1-hexanol, 96%, Thermo Scientific Chemicals

Catalog number: B21965.06
5 g, Each
Thermo Scientific Chemicals

3-Mercapto-1-hexanol, 96%, Thermo Scientific Chemicals

Catalog number: B21965.06
5 g, Each
Quantity
Catalog number: B21965.06
also known as B21965-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51755-83-0
IUPAC Name
3-sulfanylhexan-1-ol
Molecular Formula
C6H14OS
InChI Key
TYZFMFVWHZKYSE-UHFFFAOYNA-N
SMILES
CCCC(S)CCO
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥95.0%
Refractive Index
1.4775-1.4835 @ 20?C

Description

3-mercapto-1-hexanol (3MH) are compounds potentially responsible for the development of blackcurrant aroma in red wine. 3-mercapto-1-hexanol was the most important odorant of Grenache rosé wine was with a deep impact on the wine fruity and citric notes. 3-Mercaptohexanol is an thiol compound found in various grape juices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-mercapto-1-hexanol (3MH) are compounds potentially responsible for the development of blackcurrant aroma in red wine. 3-mercapto-1-hexanol was the most important odorant of Grenache rosé wine was with a deep impact on the wine fruity and citric notes. 3-Mercaptohexanol is an thiol compound found in various grape juices.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text