Chemical Identifiers
CAS73630-07-6
IUPAC Nameethyl 2-{[2-(2-{2-[bis(2-ethoxy-2-oxoethyl)amino]phenoxy}ethoxy)phenyl](2-ethoxy-2-oxoethyl)amino}acetate
Molecular FormulaC30H40N2O10
InChI KeyOLXCPQDFHUCXBA-UHFFFAOYSA-N
SMILESCCOC(=O)CN(CC(=O)OCC)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(=O)OCC)CC(=O)OCC
View more