4-Allyl-2,6-dimethoxyphenol is used as a flavoring agent or adjuvant. Also used to produce 2,6-Dimethoxy-4-propyl-phenol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Allyl-2,6-dimethoxyphenol is used as a flavoring agent or adjuvant. Also used to produce 2,6-Dimethoxy-4-propyl-phenol.
Solubility
Soluble in fats. Insoluble in water.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only
Atsuko Nishiyama; Hideo Eto; Yukimasa Terada; Masanobu Iguchi; Shosuke Yamamura. Anodic Oxidation Of 4-Allyl-2, 6-Dimethoxyphenol And Related Compounds : Syntheses Of Asatone And Related Neolignans. Chemical And Pharmaceutical Bulletin. 1983, 31 (8), 2820-2833.