4-Allyl-2,6-dimethoxyphenol, 98%, Thermo Scientific Chemicals
4-Allyl-2,6-dimethoxyphenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Allyl-2,6-dimethoxyphenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number B21981.06
also known as B21981-06
Price (USD)/ Each
126.65
Online exclusive
141.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
126.65
Online exclusive
141.00 
Save 14.35 (10%)
Add to cart
4-Allyl-2,6-dimethoxyphenol, 98%, Thermo Scientific Chemicals
Catalog numberB21981.06
Price (USD)/ Each
126.65
Online exclusive
141.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS6627-88-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1,5460-1.5500 @ 20?C
4-Allyl-2,6-dimethoxyphenol is used as a flavoring agent or adjuvant. Also used to produce 2,6-Dimethoxy-4-propyl-phenol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Allyl-2,6-dimethoxyphenol is used as a flavoring agent or adjuvant. Also used to produce 2,6-Dimethoxy-4-propyl-phenol.

Solubility
Soluble in fats. Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only
Atsuko Nishiyama; Hideo Eto; Yukimasa Terada; Masanobu Iguchi; Shosuke Yamamura. Anodic Oxidation Of 4-Allyl-2, 6-Dimethoxyphenol And Related Compounds : Syntheses Of Asatone And Related Neolignans. Chemical And Pharmaceutical Bulletin. 1983, 31 (8), 2820-2833.