Filter aid, Celite Standard Super-cel is used to maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Filter aid, Celite Standard Super-cel is used to maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization.
Solubility
Insoluble in water.
Notes
Incompatible with acids.
RUO – Research Use Only
General References:
- Farmani, J. Modeling of solid fat content of chemically interesterified fully hydrogenated soybean oil and canola oil blends as a function of temperature and saturated fatty acids. Journal of Food Measurement and Characterization 2015, 9 (3), 281-289.
- Garderen, N. V.; Otal, E. H.; Aneziris, C. G.; Graule, T.; Clemens, F. J. Influence of porous substrate on copper based oxygen carrier efficiency for chemical-looping combustion. Microporous Mesoporous Mater. 2014, 190, 362-370.