2-Amino-3,5-dichloropyridine, 98%, Thermo Scientific Chemicals
2-Amino-3,5-dichloropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-3,5-dichloropyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B21984.14
also known as B21984-14
Price (USD)/ Each
123.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
123.00
Add to cart
2-Amino-3,5-dichloropyridine, 98%, Thermo Scientific Chemicals
Catalog numberB21984.14
Price (USD)/ Each
123.00
-
Add to cart
Chemical Identifiers
CAS4214-74-8
IUPAC Name3,5-dichloropyridin-2-amine
Molecular FormulaC5H4Cl2N2
InChI KeyOCWBGKZFOYMCCN-UHFFFAOYSA-N
SMILESNC1=NC=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to Pale brown
Assay (GC)≥97.5%
FormCrystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)78-83?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only