Thermo Scientific Chemicals

2-Amino-3,5-dichloropyridine, 98%, Thermo Scientific Chemicals

Catalog number: B21984.22
100 g, Each
Thermo Scientific Chemicals

2-Amino-3,5-dichloropyridine, 98%, Thermo Scientific Chemicals

Catalog number: B21984.22
100 g, Each
Quantity
Catalog number: B21984.22
also known as B21984-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4214-74-8
IUPAC Name
3,5-dichloropyridin-2-amine
Molecular Formula
C5H4Cl2N2
InChI Key
OCWBGKZFOYMCCN-UHFFFAOYSA-N
SMILES
NC1=NC=C(Cl)C=C1Cl
Appearance (Color)
White to Pale brown
Assay (GC)
≥97.5%
Form
Crystalline powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
78-83?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text