Thermo Scientific Chemicals

2,4,5-Triamino-6-hydroxypyrimidine sulfate, 94%, Thermo Scientific Chemicals

Catalog number: B21989.14
25 g, Each
Thermo Scientific Chemicals

2,4,5-Triamino-6-hydroxypyrimidine sulfate, 94%, Thermo Scientific Chemicals

Catalog number: B21989.14
25 g, Each
Quantity
Catalog number: B21989.14
also known as B21989-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
35011-47-3
IUPAC Name
2,5,6-triamino-1,4-dihydropyrimidin-4-one; sulfuric acid
Molecular Formula
C4H9N5O5S
InChI Key
RSKNEEODWFLVFF-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.NC1=NC(=O)C(N)=C(N)N1
Form
Powder
Acid-base back titration
≥92.0 to ≤108.0%
Identification (FTIR)
Conforms
Appearance (Color)
Pale yellow to yellow or pale brown
Assay (HPLC)
≥92.0%

Description

2,4,5-Triamino-6-hydroxypyrimidine sulfate used as an intermediate for synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,5-Triamino-6-hydroxypyrimidine sulfate used as an intermediate for synthetic organic chemistry.

Solubility
Soluble in DMSO.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text