Thermo Scientific Chemicals

Polyethylene glycol 400, Thermo Scientific Chemicals

Catalog number: B21992.0B
1000 g, Each
Thermo Scientific Chemicals

Polyethylene glycol 400, Thermo Scientific Chemicals

Catalog number: B21992.0B
1000 g, Each
Quantity
Catalog number: B21992.0B
also known as B21992-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25322-68-3
Molecular Formula
(C2H4O)n
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES
[H]OCCO
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
pH
4.5-7.0 (10%, demin water)
Form
Viscous liquid
Hydroxyl Value
267- 295 mg KOH/g

Description

Polyethylene glycol is used in a variety of pharmaceutical formulations. It finds application in Hewlett-Packard designjet printers as an ink solvent and a lubricant for the print heads. It is an important raw material for the manufacture of polyuretnae, PEG salts of lauric, oleic and stearic acids and latex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text