Polyethylene glycol 400, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polyethylene glycol 400, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number B21992.30
also known as B21992-30
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
41.70 
Save 6.05 (15%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
41.70 
Save 6.05 (15%)
Add to cart
Polyethylene glycol 400, Thermo Scientific Chemicals
Catalog numberB21992.30
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
41.70 
Save 6.05 (15%)
-
Add to cart
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
pH4.5-7.0 (10%, demin water)
FormViscous liquid
Hydroxyl Value267- 295 mg KOH/g
View more
Polyethylene glycol is used in a variety of pharmaceutical formulations. It finds application in Hewlett-Packard designjet printers as an ink solvent and a lubricant for the print heads. It is an important raw material for the manufacture of polyuretnae, PEG salts of lauric, oleic and stearic acids and latex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only