Thermo Scientific Chemicals

Methyl 2,3-dichloropropionate, 98%, Thermo Scientific Chemicals

Catalog number: B21993.14
25 g, Each
Thermo Scientific Chemicals

Methyl 2,3-dichloropropionate, 98%, Thermo Scientific Chemicals

Catalog number: B21993.14
25 g, Each
Quantity
Catalog number: B21993.14
also known as B21993-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3674-09-7
IUPAC Name
methyl 2,3-dichloropropanoate
Molecular Formula
C4H6Cl2O2
InChI Key
OFHMODDLBXETIK-UHFFFAOYNA-N
SMILES
COC(=O)C(Cl)CCl
Form
Liquid
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Refractive Index
1.4515-1.4555 @ 20?C

Description

Methyl 2,3-dichloropropionate can be used to produce 2-chloro-acrylic acid methyl ester at Heating. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2,3-dichloropropionate can be used to produce 2-chloro-acrylic acid methyl ester at Heating. It is used as pharmaceutical intermediate.

Solubility
Soluble in ether, acetone, ethanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text