Thermo Scientific Chemicals

4-Chloro-2,6-dimethylaniline, 96%, Thermo Scientific Chemicals

Catalog number: B21996.06
5 g, Each
Thermo Scientific Chemicals

4-Chloro-2,6-dimethylaniline, 96%, Thermo Scientific Chemicals

Catalog number: B21996.06
5 g, Each
Quantity
Catalog number: B21996.06
also known as B21996-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24596-18-7
IUPAC Name
4-chloro-2,6-dimethylaniline
Molecular Formula
C8H10ClN
InChI Key
JCLZLZKUISPXDC-UHFFFAOYSA-N
SMILES
CC1=CC(Cl)=CC(C)=C1N
Appearance (Color)
White to pale cream to pale brown to pale gray to gray or pink
Assay (GC)
≥95.0%
Form
Crystals or powder or crystalline powder

Description

4-chloro-2,6-dimethylaniline was chosen as a substrate of medium in palladium-catalyzed cyanations of aryl chlorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-chloro-2,6-dimethylaniline was chosen as a substrate of medium in palladium-catalyzed cyanations of aryl chlorides.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text