1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%, Thermo Scientific Chemicals
1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B22003.22
also known as B22003-22
Price (USD)/ Each
275.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
275.00
Add to cart
1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%, Thermo Scientific Chemicals
Catalog numberB22003.22
Price (USD)/ Each
275.00
-
Add to cart
Chemical Identifiers
CAS52431-78-4
IUPAC Name1-(4-hydroxyphenyl)-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
Molecular FormulaC7H6N4OS
InChI KeyMOXZSKYLLSPATM-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)N1NN=NC1=S
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤5.0% (105?C/constant weight)
FormCrystals, crystalline powder or pieces
Assay (HPLC)≥95.0%
Solution Test25 mg/ml in Methanol: clear colorless to pale yellow or pink to purple solution
Appearance (Color)White to cream to purple
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only