Thermo Scientific Chemicals

trans-2,3-Epoxybutane, 97%, Thermo Scientific Chemicals

Catalog number: B22005.06
5 g, Each
Thermo Scientific Chemicals

trans-2,3-Epoxybutane, 97%, Thermo Scientific Chemicals

Catalog number: B22005.06
5 g, Each
Quantity
Catalog number: B22005.06
also known as B22005-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21490-63-1
IUPAC Name
(2S,3S)-2,3-dimethyloxirane
Molecular Formula
C4H8O
InChI Key
PQXKWPLDPFFDJP-IMJSIDKUSA-N
SMILES
C[C@@H]1O[C@H]1C
Appearance (Color)
Colorless to pale yellow
Assay from Supplier's CofA
≥96.0%
Form
Liquid

Description

A higher order cuprate reacted with trans-2,3-epoxybutane a high yield both counted on the epoxide (96%) and on the haloalkane (85%). Enolate derived from trans-2,3-Epoxybutan has chiral recognition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A higher order cuprate reacted with trans-2,3-epoxybutane a high yield both counted on the epoxide (96%) and on the haloalkane (85%). Enolate derived from trans-2,3-Epoxybutan has chiral recognition.

Solubility
Soluble in water(95g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text